missing translation for 'onlineSavingsMsg'
Learn More

Hemokinin 1, Human, Thermo Scientific™

For research use

£170.00 - £252.00

Specifications

Formulation Lyophilized powder
Storage Requirements -30° to -10°C
Regulatory Status RUO
Product Type Hemokinin 1, Human
Recombinant Native
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15416819
SDS View Documents Promotion Details
Thermo Scientific Alfa Aesar
J65288.#0
5mg This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
15426819
SDS View Documents Promotion Details
Thermo Scientific Alfa Aesar
J65288.MA
10mg This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
Description

Description

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Specifications

Lyophilized powder
RUO
Native
-30° to -10°C
Hemokinin 1, Human
Lyophilized
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only