missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
Hemoglobin zeta Monoclonal antibody specifically detects Hemoglobin zeta in Human samples. It is validated for Western Blot, Immunofluorescence
Specifications
Specifications
| Antigen | Hemoglobin zeta |
| Applications | Western Blot, Immunofluorescence |
| Classification | Monoclonal |
| Clone | GT1544 |
| Conjugate | Unconjugated |
| Formulation | PBS |
| Gene Alias | HBAZ, HBZ2, hemoglobin subunit zeta, Hemoglobin zeta chain, hemoglobin, zeta, zeta-globin |
| Host Species | Mouse |
| Immunogen | Recombinant protein encompassing a sequence within the center region of human Hemoglobin zeta. The exact sequence is proprietary. |
| Purification Method | Protein A purified |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?