missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Hemoglobin Polyclonal Antibody, Invitrogen™
GREENER_CHOICE

Rabbit Polyclonal Antibody

£161.00 - £304.00

Specifications

Antigen Hemoglobin
Concentration 0.5 mg/mL
Applications Immunohistochemistry (Paraffin), Western Blot, Western Blot
Classification Polyclonal
Conjugate Unconjugated
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
17940213
GREENER_CHOICE
View Documents
Invitrogen™
PA5145321
200 μL
£304.00
200µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
17940833
View Documents
Invitrogen™
PA5145322
20 μL
£161.00
20µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

The human alpha globin gene cluster located on chromosome 16 spans about 30 kb and includes seven loci: 5'- zeta - pseudozeta - mu - pseudoalpha-1 - alpha-2 - alpha-1 - theta - 3'. The alpha-2 (HBA2) and alpha-1 (HBA1) coding sequences are identical. These genes differ slightly over the 5' untranslated regions and the introns, but they differ significantly over the 3' untranslated regions. Two alpha chains plus two beta chains constitute HbA, which in normal adult life comprises about 97% of the total hemoglobin; alpha chains combine with delta chains to constitute HbA-2, which with HbF (fetal hemoglobin) makes up the remaining 3% of adult hemoglobin. Alpha thalassemias result from deletions of each of the alpha genes as well as deletions of both HBA2 and HBA1; some nondeletion alpha thalassemias have also been reported.
Specifications

Specifications

Hemoglobin
Immunohistochemistry (Paraffin), Western Blot, Western Blot
Unconjugated
Rabbit
Bovine, Human
P01966, P02070, P68871, P69905
100140149, 280813, 3039, 3043, 512439
Native Bovine HB protein (Accession #P01966 & P02070).
Primary
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
0.5 mg/mL
Polyclonal
Liquid
RUO
PBS with 50% glycerol and 0.05% Proclin 300; pH 7.4
alpha one globin; alpha-1 globin; alpha-1-globin; alpha-2 globin chain; Alpha-globin; ALPHA-GLOBIN GENE; delta globin; Haemoglobin; Hb; HBA1; HbA1c; HBA2; HBA-T3; HBH; hemoglobin alpha 1 globin chain; Hemoglobin alpha chain; Hemoglobin subunit alpha; hemoglobin subunit alpha 1; hemoglobin, alpha 1
HBA, HBA1, HBB
IgG
Antigen affinity chromatography, Protein A
HBB
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.