missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Hemoglobin gamma A Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA577998
This item is not returnable.
View return policy
Description
Positive Control: K562 Predicted Reactivity: Chimpanzee (99%) Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
The gamma globin genes (HBG1 and HBG2) are normally expressed in the fetal liver, spleen and bone marrow. Two gamma chains together with two alpha chains constitute fetal hemoglobin (HbF) which is normally replaced by adult hemoglobin (HbA) at birth. In some beta-thalassemias and related conditions, gamma chain production continues into adulthood. The two types of gamma chains differ at residue 136 where glycine is found in the G-gamma product (HBG2) and alanine is found in the A-gamma product (HBG1). The former is predominant at birth. The order of the genes in the beta-globin cluster is: 5'-epsilon - gamma-G - gamma-A - delta - beta-3'.
Specifications
| Hemoglobin gamma A | |
| Polyclonal | |
| Unconjugated | |
| Hbg1 | |
| A-gamma globin; epsilon 3 globin; gamma A hemoglobin; gamma globin; gamma-1-globin; Hb F Agamma; Hbb-y; Hbe1; Hbg1; HBGA; HBGR; HBG-T2; Hemoglobin gamma-1 chain; hemoglobin gamma-a chain; hemoglobin subunit gamma 1; hemoglobin subunit gamma-1; hemoglobin Y beta-like embryonic chain; hemoglobin Y, beta-like embryonic chain; hemoglobin, epsilon 1; hemoglobin, gamma A; hemoglobin, gamma, regulator of; HSGGL1; PRO2979 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 3047, 94164 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot | |
| 1 mg/mL | |
| PBS with 1% BSA, 20% glycerol and 0.01% thimerosal; pH 7 | |
| P69891 | |
| Hbg1 | |
| Full length human Hemoglobin gamma A Recombinant protein. | |
| 100 μL | |
| Primary | |
| Human, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction