missing translation for 'onlineSavingsMsg'
Learn More

Hemogard™ Lavender Closure Blood Tube

Product Code. 12316949
Change view
Click to view available options
Capacity (Metric):
3 mL
4 mL
Unit Size:
Pack of 100
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) Unit Size
12316949 3 mL Pack of 100
12326949 4 mL Pack of 100
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 12316949 Supplier Brand Not Defined Supplier No. VS367838
 Restricted Item
Estimated Shipment: 01-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

Product Type Blood Collection Tube
Additive K2 EDTA
Closure Type Hemogard
Capacity (Metric) 3 mL
Closure Color Lavender
Height (Metric) 75 mm
Diameter (Metric) Outer 13 mm
compliance-icons
Product Identifier
  • TUBE BLOOD EDTA / PLUS SPRAY COAT (K2) BL hemogard closure colour code lavender 13 x 75mm
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
Hazard Statement
  • H302-Harmful if swallowed.
Precautionary Statement
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P405-Store locked up.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contain: Dipotassium dihydrogen ethylenediaminetetraacetate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.