missing translation for 'onlineSavingsMsg'
Learn More

Hemin (porcine), 97+%

Product Code. 11409103
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11409103 5 g 5g
11499093 1 g 1g
11419103 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11409103 Supplier Thermo Scientific Alfa Aesar Supplier No. A11165.06
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 16009-13-5 | C34H32FeN4O4 | 616.50 g/mol

Hemin is used in cellular protection and control mechanism. It stimulates the synthesis of globulin. It is also used to study the bacterial strains and growth conditions, in the short-term colony assays and cell transfections. Further, it is used to treat the symptoms of various porphyrias. In addition, it is used in the preparation of agar medium for the bacteria culture.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Hemin is used in cellular protection and control mechanism. It stimulates the synthesis of globulin. It is also used to study the bacterial strains and growth conditions, in the short-term colony assays and cell transfections. Further, it is used to treat the symptoms of various porphyrias. In addition, it is used in the preparation of agar medium for the bacteria culture.

Solubility
Soluble in dilute ammonia, trimethylamine, p-toluidine and dimethylaniline. Slightly soluble in alcohol. Insoluble in water

Notes
Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 16009-13-5
Molecular Formula C34H32ClFeN4O4
Molecular Weight (g/mol) 651.95
MDL Number MFCD00010726
InChI Key BTIJJDXEELBZFS-HXFTUNQESA-K
Synonym chlorohemin, hemin chloride, protoferriheme, ferriheme, ferriprotoporphyrin ix chloride, ferriprotoporphyrin ix, panhematin, protohemin, protohemin ix, ferriprotoporphyrin
PubChem CID 131675604
IUPAC Name 3-[(1Z,6Z,12Z,17Z)-5-(2-carboxyethyl)-22-chloro-15,20-diethenyl-4,10,14,19-tetramethyl-21,23,24,25-tetraaza-22-ferrahexacyclo[9.9.3.1³,⁶.1¹³,¹⁶.0⁸,²³.0¹⁸,²¹]pentacosa-1,3(25),4,6,8,10,12,14,16(24),17,19-undecaen-9-yl]propanoic acid
SMILES CC1=C(CCC(O)=O)\C2=C\C3=C(CCC(O)=O)C(C)=C4\C=C5/N=C(/C=C6\N([Fe](Cl)N34)\C(=C/C1=N2)C(C=C)=C6C)C(C=C)=C5C

Specifications

Melting Point >300°C
Quantity 5 g
Beilstein 5229914
Merck Index 14,4644
Solubility Information Soluble in dilute ammonia,trimethylamine,p-toluidine and dimethylaniline. Slightly soluble in alcohol. Insoluble in water
Formula Weight 651.95
Percent Purity ≥98%
Chemical Name or Material Hemin (porcine)
Product Identifier
  • Hemin (porcine)
Supplemental information
  • MIXTURE LIST-Contain: Hemin

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.