missing translation for 'onlineSavingsMsg'
Learn More

Hemin, MedChemExpress

Product Code. 16481809
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16481809 1 g 1g
16480228 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16481809 Supplier Medchem Express Supplier No. HY19424
 Restricted Item
Estimated Shipment: 14-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Hemin is an iron-containing porphyrin. Hemin is an Heme oxygenase (HO)-1 inducer.

Hemin is an iron-containing porphyrin. Hemin is an Heme oxygenase (HO)-1 inducer.

Specifications

Chemical Name or Material Hemin
CAS 16009-13-5
Color Black
Purity Grade Notes Research
Molecular Formula C34H32ClFeN4O4
Quantity 1 g
Synonym Hemin chloride
Solubility Information DMSO : 17.33 mg/mL (26.58 mM; Need ultrasonic) ∣NH4OH : 2.5 mg/mL (3.83 mM; ultrasonic and adjust pH to 11 with H2O) ∣H2O : 0.1 mg/mL (0.15 mM; Need ultrasonic) ∣0.1 M NaOH : < 1 mg/mL (ultrasonic;warming;adjust pH to 12 with NaOH;heat to 60°C) (insoluble)
SMILES [Cl-][Fe+3]123[N-]4C5=C(CCC([O-])=O)C(C)=C4C=C(C(C=C)=C6C)[N]1=C6C=C(C(C=C)=C7C)[N-]2C7=CC(C(C)=C8CCC([O-])=O)=[N]3C8=C5.[H+].[H+]
Molecular Weight (g/mol) 651.94
Formula Weight 651.94
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-immunoregulation
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.