missing translation for 'onlineSavingsMsg'
Learn More

Hemin, MedChemExpress

Product Code. 16480228
Encompass_Preferred
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
This item is not returnable. View return policy

Product Code. 16480228

Brand: Medchem Express HY19424

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Hemin is an iron-containing porphyrin. Hemin is an Heme oxygenase (HO)-1 inducer.

Hemin is an iron-containing porphyrin. Hemin is an Heme oxygenase (HO)-1 inducer.

Specifications

Chemical Name or Material Hemin
CAS 16009-13-5
Color Black
Purity Grade Notes Research
Molecular Formula C34H32ClFeN4O4
Quantity 5 g
Synonym Hemin chloride
Solubility Information DMSO : 17.33 mg/mL (26.58 mM; Need ultrasonic) ∣NH4OH : 2.5 mg/mL (3.83 mM; ultrasonic and adjust pH to 11 with H2O) ∣H2O : 0.1 mg/mL (0.15 mM; Need ultrasonic) ∣0.1 M NaOH : < 1 mg/mL (ultrasonic;warming;adjust pH to 12 with NaOH;heat to 60°C) (insoluble)
SMILES [Cl-][Fe+3]123[N-]4C5=C(CCC([O-])=O)C(C)=C4C=C(C(C=C)=C6C)[N]1=C6C=C(C(C=C)=C7C)[N-]2C7=CC(C(C)=C8CCC([O-])=O)=[N]3C8=C5.[H+].[H+]
Molecular Weight (g/mol) 651.94
Formula Weight 651.94
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-immunoregulation
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.