missing translation for 'onlineSavingsMsg'
Learn More

Hemin Chloride (Technical grade), TRC

Product Code. 30401883
Change view
Click to view available options
Quantity:
1 g
5 g
10 g
Unit Size:
1g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30401883 10 g -
30408459 5 g -
30398701 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30401883 Supplier Toronto Research Chemicals Supplier No. TRCH24564810G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 16009-13-5
Molecular Formula C34H32ClFeN4O4
Molecular Weight (g/mol) 651.94
Synonym 1,3,5,8-Tetramethyl-2,4-divinylporphine-6,7-dipropionic Acid ferrichloride Chloro[dihydrogen 3,7,12,17-Tetramethyl-8,13-divinyl-2,18-porphinedipropionato(2-)]iron, Chlorohemin, Chloroprotoferriheme, Chloroprotohemin, Ferric Hemin, Ferriheme, Ferriheme Chloride, Ferriporphyrin Chloride, Ferriprotoporphyrin, Ferriprotoporphyrin IX, Ferriprotoporphyrin IX Chloride, Hemin, Hemin IX, Hemin Chloride, Iron(III) Protoporphyrin Chloride, KJB 003, Panhematin, Protoferriheme, Protohemin, Protohemin Chloride, Teichmann's Crystals
SMILES [Cl-][Fe+3]123N4=C5C=C6C(=C(C=C)C(=CC7=N1C(=CC8=C(CCC(=O)O)C(=C(C=C4C(=C5C=C)C)[N-]28)C)C(=C7C)CCC(=O)O)[N-]36)C

Specifications

Quantity 10 g
InChI Formula InChI=1S/C34H34N4O4.ClH.Fe/c1-7-21-17(3)25-13-26-19(5)23(9-11-33(39)40)31(37-26)16-32-24(10-12-34(41)42)20(6)28(38-32)15-30-22(8-2)18(4)27(36-30)14-29(21)35-25;;/h7-8,13-16H,1-2,9-12H2,3-6H3,(H4,35,36,37,38,39,40,41,42);1H;/q;;+3/p-3
Formula Weight 651.1462
Chemical Name or Material Hemin Chloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.