missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ HEMGN Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA554036
This item is not returnable.
View return policy
Description
Immunogen sequence: VISVIQDHPF KMYQDMAKRE DLAPKMCQEA AVPKILPCPT SEDTADLAGC SLQAYPKPDV PKGYILDTDQ NPAEPEEYNE TDQGI Highest antigen sequence identity to the following orthologs: Mouse - 41%, Rat - 34%.
HEMGN regulates the proliferation and differentiation of hematopoietic cells. Overexpression of HEMGN block the TPA-induced megakaryocytic differentiation in the K562 cell model. HEMGN may also prevent cell apoptosis through the activation of the nuclear factor-kappa B (NF-kB).
Specifications
| HEMGN | |
| Polyclonal | |
| Unconjugated | |
| HEMGN | |
| 4921524M03Rik; AI317176; CT155; EDAG; EDAG-1; erythroid differentiation-associated gene protein; Hemgn; Hemogen; hemopoietic gene protein; Hgn; mNDR; NDR; negative differentiation regulator protein; nuclear protein; PRO1037; PRO1620; Unknown (protein for MGC:134079) | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 55363 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.6 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q9BXL5 | |
| HEMGN | |
| Recombinant protein corresponding to Human HEMGN. Recombinant protein control fragment (Product #RP-92300). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction