missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Heme oxygenase 2 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA577832
This item is not returnable.
View return policy
Description
2 μg/mL of PA5-77832 was sufficient for detection of HO-2 in 20 μg of Rat brain lysate by colorimetric immunoblot analysis using Goat anti-rabbit IgG:HRP as the secondary antibody. ∣Detects approximately 36kDa.
Heme oxygenase, an essential enzyme in heme catabolism, cleaves heme to form biliverdin, which is subsequently converted to bilirubin by biliverdin reductase, and carbon monoxide, a putative neurotransmitter. Heme oxygenase activity is induced by its substrate heme and by various nonheme substances. Heme oxygenase occurs as 2 isozymes, an inducible heme oxygenase-1 and a constitutive heme oxygenase-2. HMOX1 and HMOX2 belong to the heme oxygenase family. Several alternatively spliced transcript variants encoding three different isoforms have been found for this gene.
Specifications
| Heme oxygenase 2 | |
| Polyclonal | |
| Unconjugated | |
| HMOX2 | |
| heme oxygenase (decycling) 2; heme oxygenase 2; heme oxygenase-2 non-reducing isoform; HMOX 2; HMOX2; HO 2; HO2; HO-2 | |
| Rabbit | |
| Protein A | |
| RUO | |
| 15369, 3163, 79239 | |
| -20°C | |
| Liquid |
| Immunohistochemistry, Immunoprecipitation, Western Blot | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.09% sodium azide; pH 7.4 | |
| O70252, P23711, P30519 | |
| HMOX2 | |
| Rat native full-length HO-2 purified from testes | |
| 100 μg | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction