missing translation for 'onlineSavingsMsg'
Learn More

Hematoxylin (Technical Grade), TRC

Product Code. 30137300
Change view
Click to view available options
Quantity:
100 mg
250 mg
500 mg
Unit Size:
100mg
250mg
500mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30137300 250 mg 250mg
30137990 500 mg 500mg
30012035 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30137300 Supplier Toronto Research Chemicals Supplier No. TRCG377708250MG
 Restricted Item
Estimated Shipment: 12-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 517-28-2
Molecular Formula C16 H14 O6
Molecular Weight (g/mol) 302.279
Synonym Hematoxylin
IUPAC Name (6aS,11bR)-7,11b-dihydro-6H-indeno[2,1-c]chromene-3,4,6a,9,10-pentol
SMILES Oc1cc2C[C@@]3(O)COc4c(O)c(O)ccc4[C@H]3c2cc1O

Specifications

Quantity 250 mg
InChI Formula InChI=1S/C16H14O6/c17-10-2-1-8-13-9-4-12(19)11(18)3-7(9)5-16(13,21)6-22-15(8)14(10)20/h1-4,13,17-21H,5-6H2/t13-,16+/m0/s1
Formula Weight 302.079
Chemical Name or Material Haematoxylin
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.