missing translation for 'onlineSavingsMsg'
Learn More

STEMCELL Technologies HemaTox™ Myeloid Kit

Product Code. 17109961
Encompass
Click to view available options
Quantity:
1 Kit
Unit Size:
1 set
This item is not returnable. View return policy

Product Code. 17109961

Brand: STEMCELL Technologies 09704

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Serum-free medium and supplement for the measurement of drug toxicity on myeloid cells

  • Screen large numbers of novel drug compounds for their effect on the proliferation and myeloid-specific differentiation of human hematopoietic stem and progenitor cells (HSPCs)
  • Compared to using the colony-forming unit (CFU) assay, the liquid culture-based HemaTox assay is more flexible: you can add test compounds at various points throughout the culture and measure their effects on more primitive or more mature progenitor cells
  • The HemaTox Myeloid Kit promotes the proliferation of CD34+ HSPCs and their differentiation into cells expressing myeloid markers such as CD13, CD14, and CD15
  • Users can choose the readout method to quantify the response and estimate the IC50 and IC90 of each compound tested
  • Test up to 160 conditions in triplicate per kit in 5 x 96-well plates
  • This kit may be used on its own or in combination with HemaTox Erythroid Kit or HemaTox Megakaryocyte Kit to assess lineage-specific drug toxicity in parallel

Specifications

Cell Type Hematopoietic Stem and Progenitor Cells, Myeloid Cells
For Use With (Application) Cell Culture; Toxicity Assay
Quantity 1 Kit
Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.