missing translation for 'onlineSavingsMsg'
Learn More

Hematoporphyrin, MedChemExpress

Product Code. 16438446
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16438446 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16438446 Supplier Medchem Express Supplier No. HYB0754
 Restricted Item
Estimated Shipment: 18-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

MedChemExpress Hematoporphyrin (Hematoporphyrin IX), a photosensitizer, is a substrate for affinity chromatography of heme-binding proteins. Hematoporphyrin can induce apoptosis in U87 glioma cells and decrease tumor growth in vivo when exposed to red light.

Hematoporphyrin (Hematoporphyrin IX), a photosensitizer, is a substrate for affinity chromatography of heme-binding proteins. Hematoporphyrin can induce apoptosis in U87 glioma cells and decrease tumor growth in vivo when exposed to red light.

Specifications

Chemical Name or Material Hematoporphyrin
CAS 14459-29-1
Color Purple
Purity Grade Notes Research
Molecular Formula C34H38N4O6
Quantity 100 mg
Synonym Hematoporphyrin IX
Solubility Information DMSO : 150 mg/mL (250.55 mM; Need ultrasonic)
SMILES O=C(O)CCC1=C2/C=C3C(CCC(O)=O)=C(C)C(/C=C(N/4)/C(C)=C(C(O)C)C4=C\C5=N/C(C(C(O)C)=C5C)=C\C(N2)=C1C)=N/3
Molecular Weight (g/mol) 598.69
Formula Weight 598.69
Grade Research
Physical Form Solid
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.