missing translation for 'onlineSavingsMsg'
Learn More

Hematoporphyrin dihydrochloride, MedChemExpress

Product Code. 16448446
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16448446 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16448446 Supplier Medchem Express Supplier No. HYB0754A
 Restricted Item
Estimated Shipment: 15-10-2026
Add to basket
Add to basket
This item is not returnable. View return policy

MedChemExpress Hematoporphyrin dihydrochloride (Hematoporphyrin IX dihydrochloride), a photosensitizer, is a substrate for affinity chromatography of heme-binding proteins. Hematoporphyrin dihydrochloride can induce apoptosis in U87 glioma cells and decrease tumor growth in vivo when exposed to red light.

Hematoporphyrin dihydrochloride (Hematoporphyrin IX dihydrochloride), a photosensitizer, is a substrate for affinity chromatography of heme-binding proteins. Hematoporphyrin dihydrochloride can induce apoptosis in U87 glioma cells and decrease tumor growth in vivo when exposed to red light.

Specifications

Chemical Name or Material Hematoporphyrin dihydrochloride
CAS 17696-69-4
Color Deep Purple
Purity Grade Notes Research
Molecular Formula C34H40Cl2N4O6
Quantity 100 mg
Synonym Hematoporphyrin IX dihydrochloride
Solubility Information DMSO : 62.5 mg/mL (93.06 mM; Need ultrasonic) ∣H2O : < 0.1 mg/mL (ultrasonic;warming;heat to 60°C) (insoluble)
SMILES O=C(O)CCC1=C2/C=C3C(CCC(O)=O)=C(C)C(/C=C(N/4)/C(C)=C(C(O)C)C4=C\C5=N/C(C(C(O)C)=C5C)=C\C(N2)=C1C)=N/3.Cl[H].Cl[H]
Molecular Weight (g/mol) 671.61
Formula Weight 671.61
Percent Purity 92.0%
Grade Research
Physical Form Powder
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.