missing translation for 'onlineSavingsMsg'
Learn More

AHLSTROM HemaSep™ Punch Card

Product Code. 31703062
Click to view available options
Capacity (Metric):
100 μL/Sample Area
Unit Size:
Pack of 100

Product Code. 31703062

Brand: AHLSTROM 8.01154.0000.AN

Please to purchase this item. Need a web account? Register with us today!

Plasma recovery fast and simple. HemaSep™ is a rapid, easy to use and cost-effective device to achieve high yield and purity plasma from whole blood.

HemaSep™ is available in two formats and their fiber-based technology allows effective separation of different volumes of undiluted whole blood. HemaSep™ Punch card allows separation of up to 100 μL of blood. The card is suitable for either manual punching or automated punching with robotic equipment.

Key Areas Of Use Include:

  • Wellness and Biomarkers
  • Infectious disease screening for human and veterinary applications
  • Preclinical and clinical studies
  • DMPK
TRUSTED_SUSTAINABILITY

Specifications

Product Type Specimen Collection and Transport Card
Closure Included No
Graduated No
Material High-purity Fiber Blend
Shape Rectangular
Sterility Non-sterile
Capacity (Metric) 100 μL/Sample Area
Certifications/Compliance ISO 9001
Color White
For Use With (Equipment) Automatic Puncher
Includes 2 Spots
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.