missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ HEMADO
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
Description
High affinity and selective adenosine A3 receptor agonist (Ki values are 1.1, 327, 1230 and > 30,000 nM for human A3, A1, A2A and A2B receptors respectively).
Specifications
Specifications
| Quantity | 10 mg |
| Formula Weight | 361.4 |
| Purity | >99% |
| Molecular Weight (g/mol) | 361.402 |
| Chemical Name or Material | HEMADO |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction