missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HELZ2 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA539332
This item is not returnable.
View return policy
Description
The protein encoded by this gene is a nuclear transcriptional co-activator for peroxisome proliferator activated receptor alpha. The encoded protein contains a zinc finger and is a helicase that appears to be part of the peroxisome proliferator activated receptor alpha interacting complex. This gene is a member of the DNA2/NAM7 helicase gene family. Alternatively spliced transcript variants encoding different isoforms have been found for this gene. [provided by RefSeq, Jul 2008].
Specifications
| HELZ2 | |
| Polyclonal | |
| Unconjugated | |
| HELZ2 | |
| PDIP1-alpha | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 85441 | |
| -20°C | |
| Liquid |
| Western Blot | |
| 1 mg/mL | |
| Dulbecco′s PBS with 150mM NaCl, 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| Q9BYK8 | |
| HELZ2 | |
| A synthetic peptide (CYRAPRDRYRDVDEY) derived from the internal region of human PRIC285 | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction