missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ HELLS Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA564099
This item is not returnable.
View return policy
Description
Immunogen sequence: VVYAPLSKKQ EIFYTAIVNR TIANMFGSSE KETIELSPTG RPKRRTRKSI NYSKIDDFPN ELEKLISQIQ PEVDRERAVV EVNIPV Highest antigen sequence identity to the following orthologs: Mouse - 85%, Rat - 85%.
This gene encodes a lymphoid-specific helicase. Other helicases function in processes involving DNA strand separation, including replication, repair, recombination, and transcription. This protein is thought to be involved with cellular proliferation and may play a role in leukemogenesis. Alternatively spliced transcript variants have been described, but their biological validity has not been determined.
Specifications
| HELLS | |
| Polyclonal | |
| Unconjugated | |
| HELLS | |
| AI323785; E130115I21Rik; helicase, lymphoid specific; helicase, lymphoid-specific; hells; ICF4; LSH; Lymphocyte-specific helicase; lymphoid-specific helicase; Lysh; Nbla10143; PASG; Proliferation-associated SNF2-like protein; SMARCA6; SWI/SNF2-related matrix-associated actin-dependent regulator of chromatin subfamily A member 6; SWI/SNF2-related, matrix-associated, actin-dependent regulator of chromatin, subfamily A, member 6; Tbc1d12; TBC1D12: TBC1 domain family, member 12; YFK8 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 3070 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 0.2 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q9NRZ9 | |
| HELLS | |
| Recombinant protein corresponding to Human HELLS. Recombinant protein control fragment (Product #RP-103497). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction