missing translation for 'onlineSavingsMsg'
Learn More

Helipyrone A, TRC

Product Code. 30138029
Change view
Click to view available options
Quantity:
2.5 mg
10 mg
50 mg
Unit Size:
10mg
2.5mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30138029 10 mg 10mg
30134807 50 mg 50mg
30007015 2.5 mg 2.5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30138029 Supplier Toronto Research Chemicals Supplier No. TRCH23050010MG
 Restricted Item
Estimated Shipment: 26-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 29902-01-0
Molecular Formula C17 H20 O6
Molecular Weight (g/mol) 320.337
Synonym 3,3'-Methylenebis[6-ethyl-4-hydroxy-5-methyl-2H-pyran-2-one], Helipyrone;
IUPAC Name 6-ethyl-3-[(6-ethyl-4-hydroxy-5-methyl-2-oxopyran-3-yl)methyl]-4-hydroxy-5-methylpyran-2-one
SMILES CCC1=C(C)C(=C(CC2=C(O)C(=C(CC)OC2=O)C)C(=O)O1)O

Specifications

Quantity 10 mg
InChI Formula InChI=1S/C17H20O6/c1-5-12-8(3)14(18)10(16(20)22-12)7-11-15(19)9(4)13(6-2)23-17(11)21/h18-19H,5-7H2,1-4H3
Formula Weight 320.126
Chemical Name or Material Helipyrone A
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.