Learn More
Invitrogen™ HELIOS Monoclonal Antibody (22F6), PE, eBioscience™

Armenian Hamster Monoclonal Antibody
Brand: Invitrogen™ 12-9883-42
Description
Description: This 22F6 monoclonal antibody reacts with human, mouse and canine Helios. Helios is a zinc-finger transcription factor first identified for its similarity to Ikaros. It is expressed in hematopoietic stem cells in both the embryo and adult mice. While the highest expression is found in regulatory T cells, Helios is not expressed in mature B cells, dendritic cells or myeloid cells. Helios can form a homodimer as well as heterodimerize with other Ikaros family members to regulate transcription. Eight isoforms of Helios exist, some of which possess a dominant-negative function that has also been found to be upregulated in lymphoid malignancies, including T-acute lymphoblastic leukemia. 22F6 has been reported to cross react with rat Helios. Applications Reported: This 22F6 antibody has been reported for use in intracellular staining followed by flow cytometric analysis. Applications Tested: This 22F6 antibody has been pre-titrated and tested by intracellular staining and flow cytometric analysis of mouse lymph node cell using the Foxp3/Transcription Factor Buffer Set (Product # 00-5523-00) and protocol. Please see BestProtocols® Section (Staining intracellular Antigens for Flow Cytometry) for staining protocol (refer to Protocol B: One-step protocol for intracellular (nuclear) proteins). This can be used at 5 μL (0.03 μg) per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number...
This gene encodes a member of the Ikaros family of zinc-finger proteins. Three members of this protein family (Ikaros, Aiolos and Helios) are hematopoietic-specific transcription factors involved in the regulation of lymphocyte development. This protein forms homo- or hetero-dimers with other Ikaros family members, and is thought to function predominantly in early hematopoietic development. Multiple transcript variants encoding different isoforms have been found for this gene, but the biological validity of some variants has not been determined.
Specifications
| HELIOS | |
| Monoclonal | |
| 5 μL/Test | |
| PBS with BSA and 0.09% sodium azide; pH 7.2 | |
| P81183, Q9UKS7 | |
| IKZF2 | |
| Affinity chromatography | |
| RUO | |
| 22779, 22807, 488510 | |
| 4°C, store in dark, DO NOT FREEZE! | |
| Liquid |
| Flow Cytometry | |
| 22F6 | |
| PE | |
| IKZF2 | |
| A730095J18Rik; ANF1A2; Helios; IKAROS family zinc finger 2; IKAROS family zinc finger 2 (Helios); ikaros family zinc finger protein 2; Ikzf2; Zfpn1a2; zinc finger DNA binding protein Helios; zinc finger protein Helios; zinc finger protein, subfamily 1A, 2 (Helios); ZNF1A2; Znfn1a2 | |
| Armenian Hamster | |
| 100 Tests | |
| Primary | |
| Human, Mouse, Canine | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.