missing translation for 'onlineSavingsMsg'
Learn More

Helios Antibody, Alexa Fluor™ 532, Novus Biologicals™

Product Code. 18431708 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18431708 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18431708 Supplier Novus Biologicals Supplier No. NBP204050AF532

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Helios Polyclonal antibody specifically detects Helios in Human samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen Helios
Applications Western Blot
Classification Polyclonal
Conjugate Alexa Fluor 532
Dilution Western Blot
Formulation 50mM Sodium Borate
Gene Alias HELIOS, IKAROS family zinc finger 2 (Helios), Ikaros family zinc finger protein 2, MGC34330, subfamily 1A, 2 (Helios), zinc finger DNA binding protein Helios, zinc finger protein Helios, ZNF1A2, ZNFN1A2
Host Species Rabbit
Immunogen The immunogen recognized by this antibody maps to a region between residue 25 and 75 of human IKAROS Family Zinc Finger 2 (Helios) using the numbering given in entry NP_057344.2 (GeneID 22807).
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Adaptive Immunity, Immunology
Primary or Secondary Primary
Gene ID (Entrez) 22807
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.