missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Helicobacter pylori urease Recombinant Rabbit Monoclonal Antibody (RM412)
GREENER_CHOICE

Product Code. 17896751
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17896751 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17896751 Supplier Invitrogen™ Supplier No. MA544576

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Recombinant Monoclonal Antibody

This antibody reacts to human Nkx3.1.

Urease iscentral to H.pylori metabolism and virulence, is necessary for its colonization of the gastric mucosa, and is a potent immunogen that elicits a vigorous immune response. Urease is an enzyme that is used for taxonomic identification and for diagnosis and follow-up after treatment, and is a vaccine candidate.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Helicobacter pylori urease
Applications ELISA
Classification Recombinant Monoclonal
Clone RM412
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide
Host Species Rabbit
Immunogen Proteins purified from Helicobacter pylori which correspond to the full length protein.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Target Species Bacteria
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.