missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Helicobacter pylori urease B Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-42850
This item is not returnable.
View return policy
Description
Specifically detects Helicobacter pylori urease B in Bacteria, Mouse samples, and it is validated for Immunohistochemistry (Paraffin), Western Blotting
Helicobacter pylori urease B Polyclonal specifically detects Helicobacter pylori urease B in Bacteria samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
| Helicobacter pylori urease B | |
| Polyclonal | |
| Western Blot 1:50000, Immunohistochemistry 1:100-1:1000, Immunohistochemistry-Paraffin 1:100-1:1000 | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| Bacteria | |
| IgG |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| PBS (pH 7.0), 1.0% BSA and 20% Glycerol with 0.01% Thimerosal | |
| Carrier-protein conjugated synthetic peptide encompassing a sequence within the C-terminus region of Helicobacter pylori Urease B. The exact sequence is proprietary. | |
| 0.1 mL | |
| Primary | |
| Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction