missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Helicobacter pylori Polyclonal Antibody, FITC

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA173161
This item is not returnable.
View return policy
Description
The PA1-73161 antibody reacts with Helicobacter pylori. PA1-73161 has been successfully used in Immunofluorescence applications. The PA1-73161 antibody was raised against whole H. pylori cells; ATCC strain 43504.
Helicobacter Pylori (H. Pylori) is known to cause peptic ulcers and chronic gastritis in human. It is associated with duodenal ulcers and may be involved in development of adenocarcimona and low grade lymphoma of mucosa associated lymphoid tissue in the stomach. It is a microaerophilic gram-Negative bacterium, helix-shaped, and with many diverse strains.
Specifications
| Helicobacter pylori | |
| Polyclonal | |
| FITC | |
| H. pylori | |
| Whole H. pylori cells; ATCC strain 43504. | |
| 1 mL | |
| Primary | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. Store in the dark. | |
| Liquid |
| Immunocytochemistry | |
| 5 mg/mL | |
| PBS with 10mg/mL BSA and 0.1% sodium azide; pH 7.2 | |
| Rabbit | |
| Protein A | |
| RUO | |
| Bacteria | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction