missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Helicobacter Pylori Antibody (HP/1334), DyLight 350, Novus Biologicals™
Shop All Bio Techne ProductsDescription
Helicobacter Pylori Monoclonal antibody specifically detects Helicobacter Pylori in H. Pylori samples. It is validated for Flow Cytometry, ELISA, Immunofluorescence
Specifications
Specifications
| Antigen | Helicobacter Pylori |
| Applications | Flow Cytometry, ELISA, Immunofluorescence |
| Classification | Monoclonal |
| Clone | HP/1334 |
| Conjugate | DyLight 350 |
| Formulation | 50mM Sodium Borate |
| Gene Alias | H. Pylori |
| Host Species | Mouse |
| Immunogen | Total sonicate of Helicobacter pylori |
| Purification Method | Protein A or G purified |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?