missing translation for 'onlineSavingsMsg'
Learn More

Helicobacter Pylori Antibody (HP/1334), Alexa Fluor™ 350, Novus Biologicals™

Product Code. 18322576 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.10mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18322576 0.1 mL 0.10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18322576 Supplier Novus Biologicals Supplier No. NBP314092AF350

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

Helicobacter Pylori Monoclonal antibody specifically detects Helicobacter Pylori in H. Pylori samples. It is validated for Flow Cytometry, ELISA, Immunofluorescence
TRUSTED_SUSTAINABILITY

Specifications

Antigen Helicobacter Pylori
Applications Flow Cytometry, ELISA, Immunofluorescence
Classification Monoclonal
Clone HP/1334
Conjugate Alexa Fluor 350
Formulation 50mM Sodium Borate
Gene Alias H. Pylori
Host Species Mouse
Immunogen Total sonicate of Helicobacter pylori
Purification Method Protein A or G purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Target Species H. pylori
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.