missing translation for 'onlineSavingsMsg'
Learn More

Helicin, 99%, Thermo Scientific™

Product Code. 10002940 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10002940 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10002940 Supplier Thermo Scientific Acros Supplier No. 226050010
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Specifications

CAS 618-65-5
Melting Point 178°C
Color White to Yellow
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (HPLC)
Packaging Glass bottle
Molecular Formula C13H16O7
MDL Number MFCD00006589
Quantity 1 g
Beilstein 31, 223
Merck Index 15, 4663
Specific Rotation Condition −60° (20°C c=2,H2O)
Specific Rotation -60
Synonym helicin, 2-beta-d-glucopyranosyloxy benzaldehyde, unii-6n4ti8p9r5, 2-formylphenyl beta-d-glucopyranoside, salicylaldehyde beta-d-glucopyranoside, helicine, salicylaldehyde b-d-glucoside, salicylaldehyde, a-d-glucoside, salicylaldehyde-beta-d-glucoside
Solubility Information Solubility in water: soluble. Other solubilities: soluble in ethanol
InChI Key BGOFCVIGEYGEOF-UJPOAAIJSA-N
SMILES OC[C@H]1O[C@@H](OC2=CC=CC=C2C=O)[C@H](O)[C@@H](O)[C@@H]1O
IUPAC Name 2-[(2S,3R,4S,5S,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)oxan-2-yl]oxybenzaldehyde
Molecular Weight (g/mol) 284.26
PubChem CID 101799
ChEBI CHEBI:73235
Formula Weight 284.26
Percent Purity 99%
Physical Form Crystalline Powder
Chemical Name or Material Helicin, 99%
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.