Learn More
HELB Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 24700-1-AP-150UL
 View more versions of this product
Product Code. 16893185
  • £349.00 / 150µL
Description
HELB gene ontology annotations related to this gene include cellular response to DNA damage stimulus; DNA replication, synthesis of RNA primer; negative regulation of double-strand break repair via homologous recombination; regulation of DNA double-strand break processing.
Specifications
Specifications
HELB
Polyclonal
Unconjugated
HELB
DNA helicase B, hDHB, HELB, helicase (DNA) B
Rabbit
Antigen Affinity Chromatography
RUO
92797
-20°C
Liquid
Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Western Blot
0.27 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Q8NG08
HELB
HELB Fusion Protein Ag20335
150 μL