missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ HeLa Membrane Cell Lysate

Product Code. 18620961 Shop All Bio Techne Products
Click to view available options
Quantity:
0.1 mg
Unit Size:
0.1mg
This item is not returnable. View return policy

Product Code. 18620961

Brand: Novus Biologicals™ NBL126467

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

prepared from whole tissue homogenates

TRUSTED_SUSTAINABILITY

Specifications

Quantity 0.1 mg
Content And Storage Store at -80C. Avoid freeze-thaw cycles.
Research Category Autophagy, Cancer, Macroautophagy, mTOR Pathway, Ubiquitin Proteasome Pathway
Target HeLa
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.