missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ HeLa Hypoxic/ Normoxic Cell Lysate

Product Code. 18786323 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
2 Vials
Unit Size:
Pack of 2
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18786323 2 Vials Pack of 2
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18786323 Supplier Novus Biologicals™ Supplier No. NBP236452
  • ONLINE EXCLUSIVE PRICE
    £372.00
    £351.75 / Pack of 2
    Save £20.25
    5% Off
 Restricted Item
Estimated Shipment: 19-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy
The HeLa Hypoxic/ Normoxic Cell Lysate is derived from human. It has been validated for the following applications: SDS-Page.
TRUSTED_SUSTAINABILITY

Specifications

For Use With (Application) SDS-Page
Tissue Cervix
Quantity 2 Vials
Storage Buffer The protein lysate is prepared in 1X Laemmli sample buffer with BME. Boiling before loading is not necessary.
Cellular Fraction Hypoxic / Normoxic
Host Species Human
Content And Storage Aliquot and store at -20°C or -80°C. Avoid freeze-thaw cycles.

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.