missing translation for 'onlineSavingsMsg'
Learn More

HEK 293 cells Rabbit anti-Human, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Brand:  Proteintech 27347-1-AP-20UL

 View more versions of this product

Product Code. 16825585

  • £135.00 / 20µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

This antibody is designed to detect the protein of the HEK293 cells.

Human Embryonic Kidney 293 cells, also often referred to as HEK 293, 293 cells, or less precisely as HEK cells are a specific cell line originally derived from human embryonic kidney cells grown in tissue culture. HEK 293 cells were generated in early 70s by transformation of cultures of normal human embryonic kidney cells with sheared adenovirus 5 DNA in Alex Van der Eb′s laboratory in Leiden, Holland.
Specifications

Specifications

HEK 293 cells
Polyclonal
Unconjugated
293, HEK 293 cells, HEK293
Peptide
20 μL
Primary
-20°C
Liquid
Immunocytochemistry, Immunofluorescence
1.2 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Rabbit
Protein A
RUO
Human
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.