missing translation for 'onlineSavingsMsg'
Learn More

Gankyrin Antibody (1F4), Alexa Fluor™ 350, R&D Systems™

Product Code. 18444103 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18444103 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18444103 Supplier Novus Biologicals Supplier No. NBP259400AF350
 Restricted Item
Estimated Shipment: 04-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A Novus product, part of R&D Systems. Mouse Monoclonal Antibody

Gankyrin Monoclonal antibody specifically detects Gankyrin in Human samples. It is validated for Western Blot, ELISA, Immunofluorescence
TRUSTED_SUSTAINABILITY

Specifications

Antigen Gankyrin
Applications Western Blot, ELISA, Immunofluorescence
Classification Monoclonal
Clone 1F4
Conjugate Alexa Fluor 350
Formulation 50mM Sodium Borate
Gene Alias 26S proteasome non-ATPase regulatory subunit 10, 26S proteasome regulatory subunit p28, ankyrin repeat protein, dJ889N15.2, gankyrin, hepatocellular carcinoma-associated protein p28-II, p28, p28(GANK), proteasome (prosome, macropain) 26S subunit, non-ATPase, 10
Host Species Mouse
Immunogen Recombinant human Gankyrin (1-226aa) purified from E. coli
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer, Cell Biology, Tumor Suppressors, Ubiquitin Proteasome Pathway
Primary or Secondary Primary
Gene ID (Entrez) 5716
Target Species Human
Content And Storage Store at 4°C in the dark.
Product Type Antibody
Form Purified
Isotype IgG1 κ
Product Line Novus
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.