Learn More
Heidolph™ Hei-VAP Reaction Extraction Package, UK-plug
Product Code. 17333110 Shop All Heidolph Instruments Products
Change view
Click to view available options
Plug Type:
UK-plug
Unit Size:
Each
Product Code. Plug Type unitSize
17333110 UK-plug Each
1 options
Product Code. 17333110 Supplier Heidolph™ Supplier No. 5742260003
  • ONLINE EXCLUSIVE PRICE
    £9725.00
    £9553.70 / Pack of 1
    Save £171.30
    1% Off
Estimated Shipment: 04-11-2026
Add to basket
Add to basket
The Hei-VAP Reaction Extraction Package features a motor lift, vapor temperature sensor, and tubing set.
  • Hei-VAP Expert Control with motor lift and vertical glassware set G6
  • Vacuum pump Rotavac Vario Tec
  • Vapor temperature sensor
  • Tubing set
TRUSTED_SUSTAINABILITY
Specifications
Type Motor Lift Rotary Evaporator
Plug Type UK-plug