Learn More
Heidolph™ Hei-VAP Basic Package, UK-plug
Product Code. 17303060 Shop All Heidolph Instruments Products
Change view
Click to view available options
Plug Type:
UK-plug
Unit Size:
Each
Product Code. Plug Type Unit Size
17303060 UK-plug Each
1 options
Product Code. 17303060 Supplier Heidolph™ Supplier No. 5713430003
  • ONLINE EXCLUSIVE PRICE
    £5645.00
    £5542.04 / Pack of 1
    Save £102.96
    1% Off
Estimated Shipment: 24-12-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
The Hei-VAP Basic Package features a hand lift, vacuum pump, and tubing set.
  • Hei-VAP Core with hand lift and vertical glassware set G3
  • Vacuum pump Rotavac Valve Tec
  • Manual vacuum controller
  • Tubing set
TRUSTED_SUSTAINABILITY
Specifications
Type Hand Lift Rotary Evaporator
Plug Type UK-plug