Learn More
Heidolph™ Hei-VAP Automatic Evaporation plus Package, UK-plug
Product Code. 17393110 Shop All Heidolph Instruments Products
Change view
Click to view available options
Plug Type:
UK-plug
Unit Size:
Each
Product Code. Plug Type Unit Size
17393110 UK-plug Each
1 options
Product Code. 17393110 Supplier Heidolph™ Supplier No. 5743831003
  • ONLINE EXCLUSIVE PRICE
    £10360.00
    £10174.66 / Pack of 1
    Save £185.34
    1% Off
Estimated Shipment: 08-02-2027
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
The Hei-VAP Automatic Evaporation plus Package features a motor lift, Woulff bottle, condenser, vacuum pump, and tubing set.
  • Hei-VAP Expert Control with motor lift and coated vertical glassware set G3B
  • Vacuum pump Rotavac Vario Control
  • Condenser for Rotavac Vario
  • Tubing set
  • Woulff bottle
  • AUTOaccurate-sensor
TRUSTED_SUSTAINABILITY
Specifications
Type Motor Lift Rotary Evaporator
Plug Type UK-plug