Learn More
Heidolph™ Hei-Connect PT1000 Clamping System for Bath Attachments
Product Code. 15237618 Shop All Heidolph Instruments Products
Click to view available options
For Use With (Equipment):
3 to 5 L Heat-On Blocks
Unit Size:
Each
Product Code. 15237618
Brand: Heidolph™ 5096320000
Only null left
Add to basket
Add to basket
Clamping system for heating bath attachments ranging from 3 to 5 L in size.
  • Includes support rod and attachment with cable inlet
  • Provides maximum safety and stability in its vertical alignment
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Equipment) 3 to 5 L Heat-On Blocks
For Use With (Application) Vertical Support for Bath Attachments
Type Support Stand
Material Stainless Steel