missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ HEF1 Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA583453
This item is not returnable.
View return policy
Description
Immunogen sequence: PPMRQAGRPD LRPEGVYDIP PTCTKPAGKD LHVKYNCDIP GAAEPVARRH QSLSPNHPPP QLGQSVGSQN DAYDVPRGVQ FLEPPA Highest antigen sequence indentity to the following orthologs: Mouse - 72%, Rat - 70%.
HEF-1 is a multifunctional protein involved in integrin-based signaling that affects cell motility, growth, apoptosis and oncogenic transformation. The Cas family of docking proteins have been the subject of intense research because of their role in cell motility, growth, apoptosis and oncogenic transformation. These proteins are substrates of focal adhesion kinase (FAK) and the Src family of tyrosine kinases two active targets for drug development. HEF1 protein production increases levels of mRNA transcripts that encode proteins associated with motility, cell transformation and invasiveness, including several metalloproteinases, MLCK, p160ROCK and ErbBi. HEF1 overproduction also mediates apoptosis in epithelial-derived cell lines, including MCF7 and HeLa cells. Recent clinical studies at another institution have found that overexpression of BCAR1 (p130Cas), a related protein, is associated with tamoxifen resistance. This highlights the importance of studying the role of this family of proteins in cancer prognosis.
Specifications
| HEF1 | |
| Polyclonal | |
| Unconjugated | |
| NEDD9 | |
| Cas scaffolding protein family member 2; CAS2; CASL; CAS-L; cas-like docking; CASS2; Crk-associated substrate lymphocyte-specific protein; Crk-associated substrate related protein Cas-L; CRK-associated substrate-related protein; dJ49G10.2; dJ761I2.1; Enhancer of filamentation 1; Enhancer of filamentation 1 p55; HEF1; mEF1; Nedd9; NEDD-9; Neural precursor cell expressed developmentally down-regulated protein 9; neural precursor cell expressed, developmentally down-regulated 9; neural precursor cell expressed, developmentally down-regulated gene 9; p105; p130Cas-related protein; renal carcinoma antigen NY-REN-12 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 4739 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot, Immunocytochemistry | |
| 0.2 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q14511 | |
| NEDD9 | |
| Recombinant protein corresponding to Human NEDD9. Recombinant protein control fragment (Product #RP-104700). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction