missing translation for 'onlineSavingsMsg'
Learn More

Hederagenin, MedChemExpress

Product Code. 16481358
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
Unit Size:
10mg
5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16481358 5 mg 5mg
16468918 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16481358 Supplier Medchem Express Supplier No. HYN02565mg
 Restricted Item
Estimated Shipment: 21-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

MedChemExpress Hederagenin is a triterpenoid saponin. It can inhibit LPS-stimulated expression of iNOS, COX-2, and NF-(kappa)B.

Hederagenin is a triterpenoid saponin. It can inhibit LPS-stimulated expression of iNOS, COX-2, and NF-(kappa)B.

Specifications

Chemical Name or Material Hederagenin
CAS 465-99-6
Color White
Purity Grade Notes Research
Molecular Formula C30H48O4
Quantity 5 mg
Solubility Information DMSO : 50 mg/mL (105.78 mM; Need ultrasonic)
SMILES CC1(C)CC[C@@](CC[C@]2(C)C3=CC[C@@]4([H])[C@@]2(C)CC[C@]5([H])[C@]4(C)CC[C@H](O)[C@]5(CO)C)(C(O)=O)[C@@]3([H])C1
Molecular Weight (g/mol) 472.7
Formula Weight 472.7
Percent Purity 98.0%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.