missing translation for 'onlineSavingsMsg'
Learn More

Hederacoside D, MedChemExpress

Product Code. 16461358
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
Unit Size:
10mg
5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16461358 5 mg 5mg
16448918 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16461358 Supplier Medchem Express Supplier No. HYN0254
 Restricted Item
Estimated Shipment: 09-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Hederacoside D is one of the bioactive saponins from Hedera helix, and plays pivotal roles in the overall biological activity.

Hederacoside D is one of the bioactive saponins from Hedera helix, and plays pivotal roles in the overall biological activity.

Specifications

Chemical Name or Material Hederacoside D
CAS 760961-03-3
Color White
Purity Grade Notes Research
Molecular Formula C53H86O22
Quantity 5 mg
Solubility Information DMSO : 100 mg/mL (93.00 mM; Need ultrasonic)
SMILES O[C@@H]1[C@H](O)[C@@H](O)C(O[C@@H]2[C@](CO)(C)C(CC[C@]3(C)C4([H])CC=C5C3(C)CC[C@]6(C(O[C@@H]7O[C@H](COC8[C@H](O)[C@@H](O)[C@H](O[C@]9([H])O[C@@H](C)[C@H](O)[C@@H](O)[C@H]9O)[C@@H](CO)O8)[C@@H](O)[C@H](O)[C@H]7O)=O)[C@@]5([H])CC(C)(C)CC6)([H])[C@]4(C)CC2)([H])OC1
Molecular Weight (g/mol) 1075.24
Formula Weight 1075.24
Percent Purity 98.47%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-immunoregulation
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.