missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ HECA Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA559945
This item is not returnable.
View return policy
Description
Immunogen sequence: RSSRYLGEFL KNAIHLEPHK KAMAGGHVFR NAHFDYSPAG LAVHRGGHFD TPVQFLRRLD LSELLTHIPR HKLNTFHVRM EDDAQVGQGE DLRKFILAAL SASHRNVVNC ALCHRALPVF EQFPLVDGTL FLSPSRHDEI EYDVP Highest antigen sequence identity to the following orthologs: Mouse - 97%, Rat - 97%.
This gene encodes the homolog of the Drosophila headcase protein, a highly basic, cytoplasmic protein that regulates the re-entry of imaginal cells into the mitotic cycle during adult morphogenesis. In Drosophila, the encoded protein also inhibits terminal branching of neighboring cells during tracheal development.
Specifications
| HECA | |
| Polyclonal | |
| Unconjugated | |
| HECA | |
| dJ225E12.1; Gm869; H dJ225E12.1; HDC; hdc homolog, cell cycle regulator; HDCL; headcase homolog; headcase homolog (Drosophila); headcase protein homolog; HECA; hHDC; si:dkey-34f16.2 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 51696 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 0.2 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q9UBI9 | |
| HECA | |
| Recombinant protein fragment of Human HECA (Product #RP-100957) | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction