Learn More
Invitrogen™ HEC1 Recombinant Rabbit Monoclonal Antibody (24GB415)
GREENER_CHOICE
Rabbit Recombinant Monoclonal Antibody
Brand:  Invitrogen™ MA557101
 View more versions of this product
Product Code. 30276166
  • £434.00 / 100µL
Description
HEC1 Recombinant Monoclonal Antibody for Western Blot, ICC/IF, Flow
Hec1 is one of several proteins involved in spindle checkpoint signaling. This surveillance mechanism assures correct segregation of chromosomes during cell division by detecting unaligned chromosomes and causing prometaphase arrest until the proper bipolar attachment of chromosomes is achieved.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
HEC1
Recombinant Monoclonal
0.16 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
O14777
NDC80
A synthesized peptide derived from human HEC1 (aa 600-642).
100 μL
Primary
Human
Antibody
IgG
Flow Cytometry, Western Blot, Immunocytochemistry
24GB415
Unconjugated
NDC80
2610020P18Rik; AV139730; HEC; HEC1; HEC-1; HEC2; HEC4; Highly expressed in cancer protein; highly expressed in cancer, rich in leucine heptad repeats; HsHec1; hsNDC80; kinetochore associated 2; kinetochore protein Hec1; Kinetochore protein NDC80 homolog; kinetochore-associated protein 2; KNTC2; NDC80; NDC80 homolog, kinetochore complex component; NDC80 homolog, kinetochore complex component (S. cerevisiae); NDC80 kinetochore complex component; NDC80 kinetochore complex component homolog; NDC80, kinetochore complex component; putative kinetochore protein NDC80 like protein; retinoblastoma-associated protein HEC; TID3
Rabbit
Affinity chromatography
RUO
10403
-20°C