missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HEBP2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 12706-1-AP-20UL
This item is not returnable.
View return policy
Description
Can promote mitochondrial permeability transition and facilitate necrotic cell death under different types of stress conditions. Does not bind hemin.Specifications
| HEBP2 | |
| Polyclonal | |
| Unconjugated | |
| HEBP2 | |
| C6orf34, C6ORF34B, HEBP2, heme binding protein 2, KIAA1244, Placental protein 23, PP23, Protein SOUL, SOUL | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 23593, 308632, 56016 | |
| -20°C | |
| Liquid |
| Western Blot, Immunoprecipitation | |
| 0.16 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| Q9WU63, Q9Y5Z4 | |
| Hebp2 | |
| HEBP2 Fusion Protein Ag3387 | |
| 20 μL | |
| Primary | |
| Human, Rat, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction