missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SPEX CertiPrep™ Heavy Metals Testing Kit
Shop All SPEX Certiprep Products
Click to view available options
:
Heavy Metals Testing Kit, 1,000 ug/mL, for ICP-MS
Description
Claritas PPT Grade Heavy Metals Testing Kit for ICP-MS, 1,000 ug/mL (1,000 ppm), 6 each 30mL
Kit contains:
- Arsenic, 1,000μg/mL, for ICP-MS, 30mL, Matrix: 2% HNO3
- Cadmium, 1,000μg/mL, for ICP-MS, 30mL,Matrix: 2% HNO3
- Chromium, 1,000μg/mL, for ICP-MS, 30mL Matrix: 2% HNO3
- Mercury, 1,000μg/mL, for ICP-MS, 30mL, Matrix: 2% HNO3
- Lead, 1,000μg/mL, for ICP-MS, 30mL Matrix: 2% HNO3
Specifications
Specifications
| Volume (Metric) | 30 mL |
| Storage Requirements | Ambient |
| Type | Certified Reference Material |
Safety and Handling
Product Identifier
- Heavy Metals Testing Kit for ICP-MS
- Danger
- Carcinogenicity Category 1A
- Carcinogenicity Category 1B
- Carcinogenicity Category 2
- Serious eye damage/eye irritation Category 1
- Serious eye damage/eye irritation Category 2
- Reproductive toxicity Category 1A
- Reproductive toxicity Category 1B
- Reproductive toxicity Category 2
- Skin corrosion/irritation Category 1 B
- Skin corrosion/irritation Category 2
- H314-Causes severe skin burns and eye damage.
- H315-Causes skin irritation.
- H318-Causes serious eye damage.
- H319-Causes serious eye irritation.
- H350-May cause cancer.
- H351-Suspected of causing cancer.
- H360-May damage fertility or the unborn child.
- H361-Suspected of damaging fertility or the unborn child.
- P240-Ground/bond container and receiving equipment.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
- P321-Specific treatment (see on this label).
- P405-Store locked up.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
- MIXTURE LIST-Contains : nitric acid, lead oxide, arsenic, cadmium, mercury
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction