missing translation for 'onlineSavingsMsg'
Learn More

Heavy Metals, plasma standard solution, Specpure™, Ag, As, Ba, Cd, Cr, Hg, Pb, Se @ 100μg/mL

Product Code. 11346968
Click to view available options
Quantity:
100 mL
500 mL
Unit Size:
100mL
500mL
This item is not returnable. View return policy

Product Code. 11346968

Brand: Thermo Scientific Alfa Aesar 014657.AE

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Heavy Metals, plasma standard solution is used as a standard solution in analytical chemistry and atomic absorption spectroscopy. It is also used as a single-element standard solution for plasma emission spectrometry.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Heavy Metals, plasma standard solution
Name Note Ag, As, Ba, Cd, Cr, Hg, Pb, Se @ 100μg/mL
Physical Form Liquid
Concentration Matrix: 5% HNO3
MDL Number MFCD02091713
UN Number UN3264
Solubility Miscible with water
Quantity 100 mL
Grade Specpure

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.