missing translation for 'onlineSavingsMsg'
Learn More

Transparent Glass Staining Trough

Product Code. 12858735
Change view
Click to view available options
Type:
Coplin Staining Jar
Staining Jar
Staining Trough
Height (Metric):
50 mm
110 mm
Unit Size:
Each
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Type Height (Metric) unitSize
12858735 Coplin Staining Jar 110 mm Each
12868735 Staining Trough 50 mm Each
12888735 Staining Jar 50 mm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 12858735 Supplier Brand Not Defined Supplier No. 2959
Only null left
Add to basket
Edge
Add to basket

Prevent evaporation of staining liquid using the drop-on lid supplied with the Transparent Glass Staining Trough. This heavy glass staining jar is an essential microscope accessory for the laboratory.

  • Glass trough with lid
  • Holds microscope slides 76mm x 26mm
  • Two different styles

Specifications

Closure Included Closure Included
Bottom Shape Flat
Color Clear
Material Glass
Type Coplin Staining Jar
For Use With (Application) For microscopic slides
Quantity 1 Each
Height (Metric) 110 mm
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.