Learn More
Manutan Heavy duty roller bearing conveyor, 400 track width, expanded 6.5m
Description
A highly versatile conveyor system for use alone or to supplement existing installations in stores, warehouses, assembly lines and loading/unloading vehicles. Easy to position and use, simply wheel to the required location, extend to the required length and lock the castor brakes for maximum stability. Very compact when closed for easy space saving storage. So flexible it can even be positioned around awkward corners as well as being used in a straight line.
- Load capacity: 125kg
- Track width: 400mm
- Closed length: 3.0m
- Expanded length: 6.5m
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.