missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Premiere Ecoforce™ Heavy Duty Catering Cleaner
Click to view available options
Volume (Metric):
5 L
Description
- Created to cut through oil and grease build-up
- Formula leaves no residual taint or odour making it perfect for food preparation areas
- Gentle on the environment in a way that still manages to deliver outstanding results
- Can be introduced into your routine in any café, restaurant or eatery to meet the highest possible hygiene standards required
Specifications
Specifications
| Product Type | Catering Cleaner |
| Certifications/Compliance | Ecolabel European environmental standard |
| For Use With (Application) | Oils and Grease |
| Quantity | 2 Pk. |
| Volume (Metric) | 5 L |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction