missing translation for 'onlineSavingsMsg'
Learn More

HEATR2 Antibody, Novus Biologicals™

Product Code. 18326249 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.05 mg
Unit Size:
0.05mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18326249 0.05 mg 0.05mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18326249 Supplier Novus Biologicals Supplier No. H00054919B01P

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Polyclonal Antibody

HEATR2 Polyclonal antibody specifically detects HEATR2 in Human samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen HEATR2
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 1:500
Formulation PBS (pH 7.4)
Gene Accession No. AAH10850
Gene Alias FLJ20397, FLJ25564, FLJ31671, FLJ39381, HEAT repeat containing 2, HEAT repeat-containing protein 2
Host Species Mouse
Immunogen FLJ20397 (AAH10850, 1 a.a. - 223 a.a.) full-length human protein. MRLKLFSILSTVLLRATDTINSQGQFPSYLETVTKDILAPNLQWHAGRTAAAIRTAAVSCLWALTSSEVLSAEQIRDVQETLMPQVLTTLEEDSKMTRLISCRIINTFLKTSGGMTDPEKLIKIYPELLKRLDDVSNDVRMAAASTLVTWLQCVKGANAKSYYQSSVQYLYRELLVHLDDPERAIQDAILDVPGSPSPSAMIGSFLRPPHPWRTVSQLNLFSS
Purification Method Protein A purified
Quantity 0.05 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 54919
Target Species Human
Content And Storage Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.