missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ HEATR1 Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA575818
This item is not returnable.
View return policy
Description
The antibody was affinity-purified from rabbit antiserum by affinity-chromatography using epitope-specific immunogen and the purity is > 95% (by SDS-PAGE).
Ribosome biogenesis factor. Involved in nucleolar processing of pre-18S ribosomal RNA. Required for optimal pre-ribosomal RNA transcription by RNA polymerase I. [UniProt].
Specifications
| HEATR1 | |
| Polyclonal | |
| Unconjugated | |
| HEATR1 | |
| AA517551; B130016L12Rik; BAP28; BC019693; FLJ10359; HEAT repeat containing 1; HEAT repeat-containing protein 1; HEAT repeat-containing protein 1, N-terminally processed; HEATR1; Protein BAP28; protein UTP10; RGD1564769; U3 small nucleolar RNA-associated protein 10 homolog; UTP10; UTP10, small subunit (SSU) processome component, homolog | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 217995, 361262, 55127 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q9H583 | |
| HEATR1 | |
| Synthetic peptide corresponding to residues in Human HEATR1. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction